missing translation for 'onlineSavingsMsg'
Learn More

Saint-Gobain Versilon™ Plasticizer-Free Tubing, Formulation 2001

Catalog No. 14170103
Encompass
Change view
Click to view available options
Diameter (English) Inner:
1/4 in.
Diameter (English) Outer:
3/8 in.
Thickness (English):
1/16 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English) Inner Diameter (English) Outer Thickness (English)
14170103 1/4 in. 3/8 in. 1/16 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14170103 Supplier Saint-Gobain Supplier No. AE300017
Only null left

Ideal for use in peristaltic pump systems, soap and detergent dispensing, ink transfer, water purification lines, chemical transfer, and food and beverage applications

Versilon™ Plasticizer-Free Tubing, Formulation 2001 is uniquely engineered to provide flexibility, chemical resistance and extended pump life in a clear, plasticizer-/oil-free tubing product.

  • Does not contaminate fluids
  • Superior flex life in peristaltic pumps
  • Chemically resistant to a wide range of fluids
  • Crystal clear for easy visual flow monitoring
  • Meets FDA criteria for food contact
  • Will not degrade over time
  • Will not release hazardous byproducts when properly incinerated
  • Does not yield any taste
  • Helps ensure accurate results from analytical instrument tests

Order Info

All products sold in 50-foot (15.24m) lengths

Compliance

Compliant with FDA 21 CFR, 177.2600

†At 73°F (23°C).

Specifications

Certifications/Compliance Compliant with FDA 21 CFR, 177.2600
Diameter (English) Inner 1/4 in.
Diameter (Metric) Inner 6.4 mm
Diameter (English) Outer 3/8 in.
Diameter (Metric) Outer 9.5 mm
Overall Length (English) 50 ft.
Thickness (English) 1/16 in.
Thickness (Metric) 1.6 mm