missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SAG

Catalog No. AAJ65248MCR
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65248MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ65248MCR Supplier Thermo Scientific™ Supplier No. J65248MCR

 

Acts as a potent agonist of Hedgehog signalling by activating the Smoothened (Smo) protein function.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 912545-86-9
Molecular Formula C28H28ClN3OS
Molecular Weight (g/mol) 490.06
InChI Key VFSUUTYAEQOIMW-UHFFFAOYSA-N
Synonym 3-Chloro-N-[trans-4-(methylamino)cyclohexyl]-N-[[3-(4-pyridinyl)phenyl]methyl]- benzo[b]thiophene-2-carboxamide; Smoothened Agonist
IUPAC Name 3-chloro-N-[4-(methylamino)cyclohexyl]-N-{[3-(pyridin-4-yl)phenyl]methyl}-1-benzothiophene-2-carboxamide
SMILES CNC1CCC(CC1)N(CC1=CC(=CC=C1)C1=CC=NC=C1)C(=O)C1=C(Cl)C2=CC=CC=C2S1

Specifications

Quantity 1 mg
Description Orange to red
Content And Storage -30°C to -10°C
For Use With (Application) Acts as a potent agonist of Hedgehog signalling by activating the Smoothened (Smo) protein function.
Formula Weight 490.06
Sensitivity Light Sensitive
Product Type SAG
Form Oil
Recommended Storage Store at -20°C

RUO – Research Use Only