accessibility menu, dialog, popup

UserName

CH3HNNNOSCl

CAS RN 912545-86-9

IUPAC Name: 3-chloro-N-[4-(methylamino)cyclohexyl]-N-{[3-(pyridin-4-yl)phenyl]methyl}-1-benzothiophene-2-carboxamide
Synonyms: smoothened agonist
Molecular Weight (g/mol): 490.06
Molecular Formula: C28H28ClN3OS
InChi Key: VFSUUTYAEQOIMW-UHFFFAOYSA-N
SMILES: CNC1CCC(CC1)N(CC1=CC(=CC=C1)C1=CC=NC=C1)C(=O)C1=C(Cl)C2=CC=CC=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results