missing translation for 'onlineSavingsMsg'
Learn More

Safranine O, Spectrum™ Chemical

$274.68 - $14,218.50

Chemical Identifiers

CAS 477-73-6
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18613061
View Documents Promotion Details
N/A 10 g Amber Glass Bottle
Each for $274.68
Only null left
 
18613063
View Documents Promotion Details
N/A 25 g Amber Glass Bottle
Each for $468.18
Only null left
 
18613062
View Documents Promotion Details
N/A 100 g Amber Glass Bottle
Each for $1,117.54
Only null left
 
18613060
View Documents Promotion Details
N/A 1 kg Amber Glass Bottle
Each for $5,160.82
Only null left
 
18613064
View Documents Promotion Details
N/A 5 kg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

477-73-6

Specifications

477-73-6
To Pass Test
Dark Brown
1
Reagent
Safranine O
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.