missing translation for 'onlineSavingsMsg'
Learn More

Safranine O, Spectrum™ Chemical

Catalog No. 18613061
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613061 10 g Amber Glass Bottle
18613063 25 g Amber Glass Bottle
18613062 100 g Amber Glass Bottle
18613060 1 kg Amber Glass Bottle
18613064 5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18613061 Supplier Spectrum Chemical Mfg Cor Supplier No. SA11510GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 477-73-6

Specifications

CAS Max % 1
Identification To Pass Test
Grade Reagent
Packaging Amber Glass Bottle
Color Dark Brown
Quantity 10 g
Chemical Name or Material Safranine O

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.