missing translation for 'onlineSavingsMsg'
Learn More

Safranin, 1% (w/v) Aqueous, Certified, LabChem™

Catalog No. p-5388334
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC223507 125 mL Poly Bottle
LC223501 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC223507 Supplier LabChem Supplier No. LC223507
Only null left

Chemical Identifiers

CAS 477-73-6 , 7732-18-5
Molecular Formula C20H19ClN4
Molecular Weight (g/mol) 350.85
MDL Number MFCD00011759
InChI Key QRYAEWIQIBAZOJ-UHFFFAOYSA-N
Synonym basic red 2, safranine o, gossypimine, safranin, safranine t, safranin o, safranin t, safranine, tolusafranine, hidaco safranine
PubChem CID 2723800
IUPAC Name 2,7-diamino-1,8-dimethyl-5-phenyl-5λ⁵-phenazin-5-ylium chloride
SMILES [Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1

Specifications

CAS Max % 99, 1
Color Red
Density 1g/mL
Packaging Poly Bottle
Linear Formula C20H19ClN4
Quantity 125 mL
Specific Gravity 1g/mL
Solubility Information Soluble in water
Formula Weight 350.85
Grade Certified
Physical Form Liquid
Chemical Name or Material Safranin, 1% (w/v) Aqueous
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage Room Temperature