missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saflufenacil Metabolite M800H35, ≥95.0% (HPLC), MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| CAS | 1246768-31-9 |
| Linear Formula | C11H14 ClFN4O4 S |
| Synonym | N-[4-Chloro-2-fluoro-5-({[(isopropylamino)sulfonyl]amino}carbonyl)phenyl]urea; 5-[(Aminocarbonyl)amino]-2-chloro-4-fluoro-N-[[(1-methylethyl)amino]sulfonyl]benzamide |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 352.77 |
| Percent Purity | ≥95.0% (HPLC) |
| Grade | Analytical Standard |