missing translation for 'onlineSavingsMsg'
Learn More

Safinamide 98.0+%, TCI America™

Catalog No. S093525MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S093525MG 25 mg
S0935100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S093525MG Supplier TCI America Supplier No. S093525MG
Only null left

Chemical Identifiers

CAS 133865-89-1
Molecular Formula C17H19FN2O2
Molecular Weight (g/mol) 302.349
MDL Number MFCD00897036
InChI Key NEMGRZFTLSKBAP-LBPRGKRZSA-N
Synonym (S)-2-[[4-[(3-Fluorobenzyl)oxy]benzyl]amino]propionamide
PubChem CID 131682
IUPAC Name (2S)-2-[[4-[(3-fluorophenyl)methoxy]phenyl]methylamino]propanamide
SMILES CC(C(=O)N)NCC1=CC=C(C=C1)OCC2=CC(=CC=C2)F

Specifications

Melting Point 212°C
Color White
Quantity 25 mg
Formula Weight 302.35
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Safinamide
RTECSNumber TX1457385
TSCA No
Recommended Storage Refrigerator