accessibility menu, dialog, popup

UserName

CAS RN 133865-89-1

CH3(R)NHOFNH2O

CAS RN 133865-89-1

IUPAC Name: (2R)-2-[({4-[(3-fluorophenyl)methoxy]phenyl}methyl)amino]propanamide
Synonyms: safinamide
Molecular Weight (g/mol): 302.35
Molecular Formula: C17H19FN2O2
InChi Key: NEMGRZFTLSKBAP-GFCCVEGCSA-N
SMILES: C[C@@H](NCC1=CC=C(OCC2=CC=CC(F)=C2)C=C1)C(N)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results