missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Safflower Yellow, TCI America™
Specifications
Specifications
| Quantity | 25 g |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Safflower Yellow |
Safety and Handling
| TSCA | No |