accessibility menu, dialog, popup

UserName

CAS RN 36338-96-2

OH(R)O(R)(R)OH(S)OH(S)OH(S)OHOHOOHHOOOHOO(R)OH(R)O(R)OH(S)OH(S)OH(R)OHOHO(Z)(E)(Z)(E)

CAS RN 36338-96-2

IUPAC Name: (2S,6Z)-4-{[(1Z,5R)-2,5-dihydroxy-3-[(2E)-3-(4-hydroxyphenyl)prop-2-enoyl]-4,6-dioxo-5-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]cyclohex-2-en-1-ylidene]methyl}-2,5-dihydroxy-6-[(2E)-1-hydroxy-3-(4-hydroxyphenyl)prop-2-en-1-ylidene]-2-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]cyclohex-4-ene-1,3-dione
Synonyms: carthamin
Molecular Weight (g/mol): 910.79
Molecular Formula: C43H42O22
InChi Key: UZPQVEVQJJKELH-HDOBETQRSA-N
SMILES: OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)[C@]1(O)C(=O)C(\C=C2\C(O)=C(C(=O)\C=C\C3=CC=C(O)C=C3)C(=O)[C@@](O)([C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)C2=O)=C(O)/C(=C(/O)\C=C\C2=CC=C(O)C=C2)/C1=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results