missing translation for 'onlineSavingsMsg'
Learn More

Safflower oil, Carthamus tinctorius

CAS: 8001-23-8

Supplier:  Thermo Scientific Chemicals J63982AP

 View more versions of this product

Catalog No. AAJ63982AP


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Solubility
Insoluble in cold water and hot water.

Notes
Supplied as a raw oil without preservative
Specifications

Specifications

Safflower oil
8001-23-8
0.921
1.476
500 mL
Insoluble in cold water and hot water.
Carthamus tinctorius
Yellow
>113°C (235°F)
MFCD00132216
Carthamus tinctorius oil
Oil

Chemical Identifiers

8001-23-8
MFCD00132216
Carthamus tinctorius oil
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-276-5

RTECSNumber : VN2230000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only