Learn More
Accuform™ Safety Sign - STOP TO ENTER PROOF OF VACCINATION REQUIRED, Plastic
Accuform™ vaccination signs communicate health and safety policies, reinforce procedures, and identify locations
Supplier:  Accuform™ MBDX506VP
 View more versions of this product
Catalog No. 19-119-064
Only null left
Explore available promotions
Description
Description
  • Durable plastic has slight flex ideal for indoor and sheltered outdoor environments
  • Printed surface has clear UV protective overcoat
  • 50% recycled content with UV inhibitors and good impact resistance
  • Rounded corners with 0.1875 in. mounting holes for installation
  • Complies with OSHA 29 CFR 1910.145 and ANSI Z535 requirements
  • Specifications
    Specifications
    STOP TO ENTER PROOF OF VACCINATION REQUIRED
    Red on White
    Product Suggestions
    Product Suggestions