missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Accuform™ Safety Sign - SANITIZE HANDS HERE (Down Arrow), Adhesive Vinyl
Accuform™ signs alert and remind others to follow health and safety policies to prevent spread of germs and viruses
Supplier: Accuform™ MRST587VS
Description
Specifications
| Vinyl | |
| 10 in. | |
| Rectangular | |
| SANITIZE HANDS HERE |
| Self-adhesive | |
| 7 in. | |
| English | |
| White on Blue |