Learn More
Accuform™ Safety Sign - ENTRANCE ONLY, Plastic
Accuform™ signs identify/indicate potential, type and degree of hazards that may lead to accidental injury/property damage
Supplier:  Accuform™ MADM434VP
 View more versions of this product
Catalog No. 19-119-180
Only null left
Explore available promotions
Description
Description
  • Durable plastic has slight flex ideal for indoor and sheltered outdoor environments
  • Printed surface has clear UV protective overcoat
  • 50% recycled content with UV inhibitors and good impact resistance
  • Rounded corners with 0.1875 in. mounting holes for installation
  • Complies with OSHA 29 CFR 1910.145 and ANSI Z535 requirements
  • Specifications
    Specifications
    Polyethylene
    Rectangular
    ENTRANCE ONLY
    Product Suggestions
    Product Suggestions