Learn More
SOL-GUARD Safety Pull-Button Blood Collection Set
With ease, safety and comfort in mind, this product features an exclusive version of the 'Butterfly Needle', designed with sliding button that using pull force, activates in-vein needle retraction, helping prevent needlestick injury and bloodborne pathogen exposure.
Supplier:  SOL-GUARD PBH2512
 View more versions of this product
Catalog No. 22024486
Only null left
Description
Description
  • Low angle tri-bevel tip designed to improve penetration, while minimizing patient discomfort
  • Thin-walled needle for fast flow rate and to reduce the risk of blood hemolysis
  • Tactile non-slip grip to help engage the safety feature
  • 30% wider wings for improved handling during insertion and removal
  • Large, oversized arrow to easily identify the activation pull button
  • Flashback for instant and clear confirmation of proper needle placement
  • Spring mechanism for split second in-vein needle retraction
Specifications
Chemical Identifiers
Butterfly Needle
Sterile
Yes
30.5 cm
Polyurethane
Blood Collection
12 in.
Specifications
Butterfly Needle
Polyurethane
Sterile
Blood Collection
12 in.
Yes
Orange
25 G
510K, FDA, and ISO
Yes
30.5 cm
Product Suggestions
Product Suggestions