Learn More
Sol-Care™ Safety Lancets
Safety Lancet is a sterile, single use, springe-loaded lancet designed for capillary blood sampling. The lancet is precision sharpened designed for maximum comfort and optimal blood flow.
Supplier: Sol-Care™ SLA004
Description
- The needle is safely concealed in the lancet body before and after use
- Easy twist-off cap removal
- Comfortable finger recess design
- Small contact face for precise puncturing
Chemical Identifiers
| Lancet | |
| Blood Collection Tube Holder |
Specifications
| Polypropylene | |
| Blood Collection Tube Holder | |
| 28 G |
| Non-sterile | |
| 1.8 mm | |
| Lancet |