missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Safety-Labeled Wash Bottles

Description
- Easy-to-read printed bottles
- Wide mouth 53mm (2 in.) bottle for easy refilling
- Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
- Polypropylene closures
- Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Use only with solvents compatible with LDPE.
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Color | Natural |
| Material | Polyethylene |
| Closure Color | Blue |
| Mouth | Wide |
| Chemical Label | Deionized Water |
| Closure Material | Polypropylene |
| Closure Size | 53 mm |
| Autoclavable | Not Autoclavable |
| Capacity (English) | 17 oz. |
| Show More |