missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Safety Bottle Carrier

The Safety Bottle Carrier provides a safe and reliable way to transport a variety of bottled reagents in your lab.

Supplier:  Fisherbrand™ 120913F2

 View more versions of this product

Catalog No. 16985679



Only null left

Description

Description

  • Robust Design
  • Ergonomic handle prevents hand fatigue and related accidents when carrying heavier loads
  • Handle can be pressed down to either side for easier loading and unloading
  • Centering ring stabilizes bottle in the center of carrier for safer, steadier transport
  • Carriers come in four colors with designated labeling areas for easy identification
  • Made of chemical resistant thermoplastic elastomer - PP blend
Specifications

Chemical Identifiers

Not Autoclavable
Polypropylene
Cylindrical
Non-sterile
-20 to 60°C
1 Safety Bottle Carrier vessel and attachable handle
None
Bottles
1
No
Transportation of bottled regeants
.25 kg
Bottle Holder
1 x 1

Specifications

Safety Bottle Carrier, 4.5 L, Blue
Blue
Polypropylene
Cylindrical
Non-sterile
4.5 L bottles
-20 to 60°C
18.3 cm
1 Safety Bottle Carrier vessel and attachable handle
18.3 cm
26.7 cm (40.6 cm Height, Handle Extended)
18.3 cm
Not Autoclavable
Bottles
1
No
Transportation of bottled regeants
4.5 L
18.3 cm
.25 kg
Bottle Holder
None
1 x 1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions