missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Safety Bottle Carrier
The Safety Bottle Carrier provides a safe and reliable way to transport a variety of bottled reagents in your lab.
Supplier: Fisherbrand™ 120916F2
Description
- Robust Design
- Ergonomic handle prevents hand fatigue and related accidents when carrying heavier loads
- Handle can be pressed down to either side for easier loading and unloading
- Centering ring stabilizes bottle in the center of carrier for safer, steadier transport
- Carriers come in four colors with designated labeling areas for easy identification
- Made of chemical resistant thermoplastic elastomer - PP blend
Chemical Identifiers
| Not Autoclavable | |
| Polypropylene | |
| Cylindrical | |
| Non-sterile | |
| -20 to 60°C | |
| 1 Safety Bottle Carrier vessel and attachable handle | |
| None |
| Bottles | |
| 1 | |
| No | |
| Transportation of bottled regeants | |
| .25 kg | |
| Bottle Holder | |
| 1 x 1 |
Specifications
| Safety Bottle Carrier, 1.5 L, Black | |
| Black | |
| Polypropylene | |
| Cylindrical | |
| Non-sterile | |
| 1.5 L bottles | |
| -20 to 60°C | |
| 11.4 cm | |
| 1 Safety Bottle Carrier vessel and attachable handle | |
| 11.4 cm | |
| 16.5 cm (25.4 cm Height, Handle Extended) | |
| 11.4 cm |
| Not Autoclavable | |
| Bottles | |
| 1 | |
| No | |
| Transportation of bottled regeants | |
| 1.5 L | |
| 11.4 cm | |
| .25 kg | |
| Bottle Holder | |
| None | |
| 1 x 1 |