missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sol-Care™ Safety Blood Collection Needle
A safety-engineered device with a sliding safety shield that clicks when activated, alerting you that the needle is safely locked.
Supplier: Sol-Care™ 110201030031
Description
- Transparent design that allows visualization of the blood flashback and flow
- Single-handed activation option that provides more flexibility to care for the patient and venipuncture site
- Sliding safety shield pushes over the needle and locks with an audible click, protecting against needlestick injury
Specifications
| Single-use | |
| 12 in. | |
| Safety Needle System | |
| 21 G | |
| Blood Collection Tube Holder | |
| 3/4 in. |
| Luer Adapter | |
| Sterile | |
| Yes | |
| Polypropylene | |
| Not Autoclavable | |
| 19.05 mm |