missing translation for 'onlineSavingsMsg'
Learn More

Sol-Care™ Safety Blood Collection Needle

A safety-engineered device with a sliding safety shield that clicks when activated, alerting you that the needle is safely locked.

Supplier:  Sol-Care™ 110201030030*

 View more versions of this product

Catalog No. 22024460

Only null left
Explore available promotions

Description

Description

  • Transparent design that allows visualization of the blood flashback and flow
  • Single-handed activation option that provides more flexibility to care for the patient and venipuncture site
  • Sliding safety shield pushes over the needle and locks with an audible click, protecting against needlestick injury
Specifications

Specifications

Single-use
7 in.
Safety Needle System
21 G
Blood Collection Tube Holder
3/4 in.
Luer Adapter
Sterile
Yes
Polypropylene
Not Autoclavable
19.05 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions