missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Medicom™ SafeTouch™ Rejuvenate™ Latex Medical Examination Glove with Textured Fingertips
Medicom™ SafeTouch™ Rejuvenate™ Latex Medical Examination Gloves with Textured Fingertips, Infused with lanolin and vitamin E.
Supplier: Medicom™ 1163A
Description
- Heavier weight, highly resistant glove.
- Made from natural rubber latex for an ergonomic fit and optimal tear, puncture and chemical resistance.
- Textured fingertips for a secure grip, even when wet.
- Skin-friendly: Infused with lanolin and vitamin E to moisturize and soften hands.
- 1000/Cs.
Specifications
No | |
Healthcare | |
Cream | |
Fully Textured | |
Powder Free | |
Non-sterile | |
Beaded |
No | |
1.5 | |
Yes | |
No | |
X-Small | |
No |