missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medicom™ SafeTouch™ Connect™ Latex Medical Examination Gloves
SafeTouch™ Connect™ Latex Medical Examination Gloves - Low protein content.
Supplier: Medicom™ 1124C
Description
- Heavier weight, highly resistant, low protein content latex glove.
- Fully textured for optimal grip.
- 1000/Cs.
Specifications
| No | |
| Healthcare | |
| Cream | |
| Fully Textured | |
| Latex | |
| Medium | |
| No |
| No | |
| 1.5 | |
| Single-use | |
| No | |
| Powder Free | |
| Non-sterile | |
| Beaded |