Learn More
Medicom™ SafeMask™ Master Series™ Earloop Face Masks
Engineered with Simply Soft™ technology for softness and a comfortable fit without compromising protection or breathability. Available in all 3 levels of ASTM protection and in a variety of vibrant colors.
Supplier:  Medicom™ 2056
 Afficher plus de versions de ce produit
Numéro de catalogue. 19-910-666
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Variety of unique and vibrant colors
  • Simply Soft Technology™ for a smooth, luxurious and comfortable fit
  • Breathability and softness
  • Level 1, L2 and L3 protection
  • Ultrasonically sealed earloops
  • Shingle pleat design prevents fluid pooling and cross-contamination
  • High-quality media ensures optimal filtration
  • Intended to be worn during medical procedures to protect the patient and the operating personnel
  • Latex free
Spécifications
Spécifications
Medical/Procedural Face Mask
Pink
Spunbond Polypropylene
Non-sterile
ASTM F2299, ASTM F1862
3-ply
≥95% (PFE), ≥95% (BFE)
Yes
0.1 μm
ASTM Level 2
Earloops
Standard
Pleated
EN 14683
Single-use
Medical procedures
50 Masks/Box, 10 Boxes/Cs.
SafeMask™ Master Series
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS