missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ SafeMask™ Master Series™ Earloop Face Masks

Catalog No. p-7163495
Change view
Click to view available options
Color:
Blue
Green
Pink
Purple
ASTM Level:
ASTM Level 1
ASTM Level 2
ASTM Level 3
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color ASTM Level
19910661 Blue ASTM Level 1
19910663 Green ASTM Level 1
19910662 Pink ASTM Level 1
19910665 Purple ASTM Level 1
19910667 Blue ASTM Level 2
19910666 Pink ASTM Level 2
19910668 Blue ASTM Level 3
19910669 Pink ASTM Level 3
19910670 Purple ASTM Level 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19910661 Supplier Medicom™ Supplier No. 2050
Only null left

Engineered with Simply Soft™ technology for softness and a comfortable fit without compromising protection or breathability. Available in all 3 levels of ASTM protection and in a variety of vibrant colors.

  • Variety of unique and vibrant colors
  • Simply Soft Technology™ for a smooth, luxurious and comfortable fit
  • Breathability and softness
  • Level 1, L2 and L3 protection
  • Ultrasonically sealed earloops
  • Shingle pleat design prevents fluid pooling and cross-contamination
  • High-quality media ensures optimal filtration
  • Intended to be worn during medical procedures to protect the patient and the operating personnel
  • Latex free

Specifications

Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 1
Color Blue
Fastening Type Earloops
Material Spunbond Polypropylene
Size Standard
Sterility Non-sterile
Style Pleated
Certifications/Compliance ASTM F2299, ASTM F1862
Certifications/Compliance (Europe) EN 14683
Construction 3-ply
Disposable Single-use
Filter Efficiency ≥95% (PFE), ≥95% (BFE)
For Use With (Application) Medical procedures
Latex Free Yes
Packaging Quantity 50 Masks/Box, 10 Boxes/Cs.
Pore Size 0.1 μm
Series SafeMask™ Master Series
Show More Show Less