Learn More
Eppendorf SafeCode Plates
For optimized sample identification in the laboratory, the use of barcodes for the identification of plates is recommended. The SafeCode system for Eppendorf Plates is based on a pre-labeled 3-level coding for safe sample identification based on barcodes.
Supplier:  Eppendorf 0030113632
 View more versions of this product
Catalog No. 14-557-228
Only null left
Explore available promotions
Description
Description
  • 3-level coding for safe sample and vessel identification
  • Codes on three sides of plates for flexible reading
  • ECC 200 error correction function enables safe code reading with up to 25 % damaged codes
  • Additional, ID-specific information on plates available via Eppendorf DataPort for documentation purposes
  • Direct import into eLabInventory and eLabJournal™ software for convenient handling
  • Import to other database systems possible
  • Manufactured from high-quality polypropylene for highest sample purity
Specifications
Specifications
Alphanumeric
8 x 12
Red
Skirted
150 μL
Non-sterile
Round, Conical Bottom (V-well)
Colorless
SBS Standard
25 Plates
PCR Plate
Polypropylene, Polycarbonate
96
Non-treated
25 Pk.
Product Suggestions
Product Suggestions