Learn More
ChemDAQ™ SafeCide™ 2.1 Peracetic Acid Monitor System - Handheld
Description
The SafeCide™ 2.1 Handheld Peracetic Acid (PAA) Monitor is a portable, real-time gas detection system designed for fast, accurate monitoring of peracetic acid vapor in disinfection and sterilization environments. Compact and lightweight, this handheld unit is ideal for spot-checking, area validation, and personnel safety in any location where PAA is used or stored. With real-time readings, customizable alarms, and data logging capabilities, the SafeCide™ 2.1 handheld monitor helps ensure a safe working environment while maintaining compliance with workplace exposure limits.
Key Features
- Portable and Easy to Use The SafeCide™ 2.1 handheld monitor fits comfortably in one hand and is ready to use within minutes. Its intuitive touchscreen interface makes it simple to read and operate, even with gloves on. Ideal for technicians, safety officers, and facility managers needing mobility and speed.
- Real-Time PAA Monitoring The device continuously measures peracetic acid vapor levels and displays instantaneous exposure concentrations, giving users immediate feedback during cleaning cycles, sterilization prep, or post-treatment ventilation.
- TWA & STEL Exposure Tracking In addition to real-time data, the monitor calculates Time-Weighted Average (TWA) and Short-Term Exposure Limit (STEL) values, allowing users to assess cumulative exposure risks in accordance with OSHA and ACGIH guidelines.
- Hot-Swappable SXP™ Sensors The system uses ChemDAQ's Sensor Exchange Program (SXP™), allowing fast, pre-calibrated sensor replacement in the field—no tools or calibration gas required. Reduce downtime and eliminate the hassle of manual calibration.
- Custom Alarms & Visual Indicators Integrated LED indicators and an audible alarm alert users when exposure exceeds configurable thresholds. Whether for low-level warnings or high-level emergencies, SafeCide™ 2.1 provides clear notifications that help prevent overexposure.
- Data Logging & Reporting Capture exposure data for later review. The handheld unit logs monitoring sessions and can upload data to a PC or ChemDAQ platform for documentation, analysis, or compliance auditing.
Peracetic acid is widely used for surface and equipment disinfection, but exposure to its vapors can be harmful. The SafeCide™ 2.1 Handheld Monitor gives safety professionals, auditors, and technicians a powerful yet portable solution for real-time exposure measurement, whenever and wherever it's needed. With ChemDAQ's proven sensing technology, intuitive design, and compliance-ready features, SafeCide™ 2.1 is a trusted tool in any workplace where PAA safety matters.
Specifications
Specifications
| Components Measured | Peracetic Acid |
| For Use With (Equipment) | ChemDAQ Gas Sensor |
| Type | Portable Gas Detection Monitor |
Purchase of this product requires and annual service contract for sensor calibration and exchange. Contact your Fisher Scientific representative to confirm your needs and provide a quote.