Learn More
Medicom™ SafeBasics™ Self-Sealing Sterilization Pouches
SafeBasics™ Self-Sealing Sterilization Pouches with dual indicators to confirm sterilization.
Supplier:  Medicom™ 99030
 View more versions of this product
Catalog No. 296367157
Only null left
Description
Description
  • Self-sealing sterilization pouches made from high-quality materials to deliver reliable sterilization processing at an affordable price.
  • Dual (internal/external) steam indicators provide reliable confirmation of sterilization.
Specifications
Chemical Identifiers
Autoclavable
Green
Paper
Self-adhesive
Single-use
Non-sterile
Specifications
Autoclavable
Green
Paper
7.5 x 13 in.
Self-adhesive
Single-use
Non-sterile
Product Suggestions
Product Suggestions