Learn More
Medicom™ SafeBasics™ Self-Sealing Sterilization Pouches
SafeBasics™ Self-Sealing Sterilization Pouches with dual indicators to confirm sterilization.
Supplier: Medicom™ 99025
Description
- Self-sealing sterilization pouches made from high-quality materials to deliver reliable sterilization processing at an affordable price.
- Dual (internal/external) steam indicators provide reliable confirmation of sterilization.
Chemical Identifiers
| Autoclavable | |
| Green | |
| Paper |
| Self-adhesive | |
| Single-use | |
| Non-sterile |
Specifications
| Autoclavable | |
| Green | |
| Paper | |
| 5.25 x 10 in. |
| Self-adhesive | |
| Single-use | |
| Non-sterile |