missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Safe-Lock Tubes 2.0 mL - Microtube

Catalog No. 054026
Change view
Click to view available options
Color:
Amber
Assorted
Blue
Clear
Green
Red
Yellow
Purity or Quality Grade:
Biopur™ (Sterile)
Eppendorf Quality
PCR Clean (Non-sterile)
Max. RCF:
22,000 x g
25,000 x g
Packaging:
100 Tubes, Individually Wrapped
500 Tubes ( 5 Bags x 100 Tubes)
500 Tubes (1 Bag x 500 Tubes)
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Purity or Quality Grade Max. RCF Packaging
054026 Assorted Eppendorf Quality 25,000 x g 500 Tubes ( 5 Bags x 100 Tubes)
0540211 Red Eppendorf Quality 25,000 x g 500 Tubes (1 Bag x 500 Tubes)
054029 Blue Eppendorf Quality 25,000 x g 500 Tubes (1 Bag x 500 Tubes)
0540212 Yellow Eppendorf Quality 25,000 x g 500 Tubes (1 Bag x 500 Tubes)
0540210 Green Eppendorf Quality 25,000 x g 500 Tubes (1 Bag x 500 Tubes)
054027 Clear Eppendorf Quality 25,000 x g 500 Tubes (1 Bag x 500 Tubes)
0540295 Clear PCR Clean (Non-sterile) 25,000 x g 500 Tubes (1 Bag x 500 Tubes)
054028 Amber PCR Clean (Non-sterile) 25,000 x g 500 Tubes (1 Bag x 500 Tubes)
0540224C Clear Biopur™ (Sterile) 22,000 x g 100 Tubes, Individually Wrapped
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 054026 Supplier Eppendorf™ Supplier No. 022363476

Eppendorf™ Safe-Lock Tubes 2.0 mL - Microtube ensures secure sample preservation. With an innovative Safe-Lock mechanism, these microtubes prevent accidental openings, providing reliability in a 2.0 mL capacity. Ideal for various applications, they combine durability, ease of use, and sample protection.

  • The Eppendorf Safe-Lock tube with a hinged lid prevents accidental opening during incubation and storage.
  • g-Safe™ technology, dependent on the Safe-Lock Tube variant (refer to the manual), ensures exceptional centrifugation stability up to 30,000 x g, preventing sample loss from tube breakage and enhancing safety when handling hazardous samples.
  • Manufactured without the use of slip agents, plasticizers, and biocides – substances known to leach from plastic consumables into the sample, negatively affecting bio-assay results.
  • Polypropylene construction imparts high resistance to chemicals, mechanical stress, and extreme temperatures.
  • Precise lid sealing minimizes evaporation rates during prolonged storage.
  • The tube features a large frosted lid and side surface for easy labeling.
  • Functionality is assured within a temperature range from -86 °C to 100 °C.
  • Autoclavable when open (121°C, 20 min.) for effective sterilization.
  • Available in lot-certified PCR Clean, Eppendorf Forensic DNA Grade, and Biopur™ quality for tested purity.

Order Info

Assorted packages include 100 each of natural, blue, green, red and yellow

Specifications

Closure Material Polypropylene
Autoclavable Autoclavable
Capacity (Metric) 2 mL
Color Assorted
Material Polypropylene
No. per Pack 500 Pk.
Packaging 500 Tubes ( 5 Bags x 100 Tubes)
Temperature Range (English) -122.8°F to +212°F
Temperature Range (Metric) -86°C to +100°C
Sterility Non-sterile
Purity or Quality Grade Eppendorf Quality
Graduated Yes
Capacity (English) 0.07 oz.
Closure Type Safe-lock Lid
Bottom Shape Round
Max. RCF 25,000 x g
Diameter (Metric) 11 mm
Length (Metric) 42 mm
Product Type Locking Snap-cap Microtube
Show More Show Less