missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Safe-Lock Tubes 2.0 mL - Microtube
Description
- The Eppendorf Safe-Lock tube with a hinged lid prevents accidental opening during incubation and storage.
- g-Safe™ technology, dependent on the Safe-Lock Tube variant (refer to the manual), ensures exceptional centrifugation stability up to 30,000 x g, preventing sample loss from tube breakage and enhancing safety when handling hazardous samples.
- Manufactured without the use of slip agents, plasticizers, and biocides – substances known to leach from plastic consumables into the sample, negatively affecting bio-assay results.
- Polypropylene construction imparts high resistance to chemicals, mechanical stress, and extreme temperatures.
- Precise lid sealing minimizes evaporation rates during prolonged storage.
- The tube features a large frosted lid and side surface for easy labeling.
- Functionality is assured within a temperature range from -86 °C to 100 °C.
- Autoclavable when open (121°C, 20 min.) for effective sterilization.
- Available in lot-certified PCR Clean, Eppendorf Forensic DNA Grade, and Biopur™ quality for tested purity.
Order Info
Assorted packages include 100 each of natural, blue, green, red and yellow
Specifications
Specifications
| Closure Material | Polypropylene |
| Autoclavable | Autoclavable |
| Capacity (Metric) | 2 mL |
| Color | Yellow |
| Material | Polypropylene |
| No. per Pack | 500 Pk. |
| Packaging | 500 Tubes (1 Bag x 500 Tubes) |
| Temperature Range (English) | -122.8°F to +212°F |
| Temperature Range (Metric) | -86°C to +100°C |
| Sterility | Non-sterile |
| Show More |