missing translation for 'onlineSavingsMsg'
Learn More

Saccharin Sodium, Dihydrate, Powder, USP, 99-101%, Spectrum™ Chemical

Catalog No. 18606121
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606121 2.5 kg Poly Bottle
18606119 100 g Poly Bottle
18606120 500 g Poly Bottle
18606122 12 kg Poly Bottle
18606124 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18606121 Supplier Spectrum Chemical Mfg Cor Supplier No. SO2002.5KG
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SO200, 6155-57-3, C7H4NNaO3S·2H2O

Spectrum™ Chemical Saccharin Sodium, Dihydrate, Powder, USP is a pharmaceutical grade substance whose most common use is as an artificial sweetener. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6155-57-3
Molecular Formula C7H8NNaO5S
Molecular Weight (g/mol) 241.19
InChI Key NAUHOSQQKJTUAX-UHFFFAOYSA-M
IUPAC Name sodium 1,3-dioxo-3H-1λ6,2-benzothiazol-1-olate dihydrate
SMILES O.O.[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 2.5 kg
Percent Purity 99 to 101%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Saccharin Sodium, Dihydrate, Powder