missing translation for 'onlineSavingsMsg'
Learn More

Saccharin, Powder, NF, 99-101%, Spectrum™ Chemical

Catalog No. 18606128
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606128 125 g Poly Bottle
18606129 500 g Poly Bottle
18606130 2.5 kg Poly Pail
18606131 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18606128 Supplier Spectrum Chemical Mfg Cor Supplier No. SA105125GM

SA105, 81-07-2, C7H5NO3S

Spectrum™ Chemical Saccharin, Powder, NF is a pharmaceutical grade substance whose most common use is as an artificial sweetener. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 81-07-2
Molecular Formula C7H5NO3S
Molecular Weight (g/mol) 183.18
InChI Key CVHZOJJKTDOEJC-UHFFFAOYSA-N
IUPAC Name 2,3-dihydro-1λ⁶,2-benzothiazole-1,1,3-trione
SMILES O=C1NS(=O)(=O)C2=CC=CC=C12

Specifications

CAS Max % 1
Ignition Residue 0.002
Loss on Drying 0.01
Melting Range 226-230°C
Packaging Poly Bottle
Quantity 125 g
Percent Purity 99 to 101%
Grade NF