missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saccharin, Powder, FCC, 98-101%, Spectrum™ Chemical
S1003, 81-07-2, C7H5NO3S
$614.64 - $4010.24
Chemical Identifiers
| CAS | 81-07-2 |
|---|---|
| Molecular Formula | C7H5NO3S |
| Molecular Weight (g/mol) | 183.18 |
| InChI Key | CVHZOJJKTDOEJC-UHFFFAOYSA-N |
| IUPAC Name | 2,3-dihydro-1λâ¶,2-benzothiazole-1,1,3-trione |
| SMILES | O=C1NS(=O)(=O)C2=CC=CC=C12 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18606125
|
Spectrum Chemical Mfg Cor
S1003125GM |
125 g | Poly Bottle |
Each for $614.64
|
|
||||
|
18606126
|
Spectrum Chemical Mfg Cor
S1003500GM |
500 g | Poly Bottle |
Each for $1,310.24
|
|
||||
|
18606127
|
Spectrum Chemical Mfg Cor
S100325KG |
2.5 kg | Poly Pail |
Each for $4,010.24
|
|
||||
Description
Spectrum™ Chemical Saccharin, Powder, FCC is used as an artificial sweetener. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 81-07-2 | |
| 183.18 | |
| 2,3-dihydro-1λâ¶,2-benzothiazole-1,1,3-trione |
| C7H5NO3S | |
| CVHZOJJKTDOEJC-UHFFFAOYSA-N | |
| O=C1NS(=O)(=O)C2=CC=CC=C12 |
Specifications
| 81-07-2 | |
| 0.002 | |
| 226-230°C | |
| C7H5NO3S | |
| CVHZOJJKTDOEJC-UHFFFAOYSA-N | |
| 2,3-dihydro-1λâ¶,2-benzothiazole-1,1,3-trione | |
| 98 to 101% |
| 1 | |
| 0.01 | |
| Poly Bottle | |
| 125 g | |
| O=C1NS(=O)(=O)C2=CC=CC=C12 | |
| 183.18 | |
| FCC |