missing translation for 'onlineSavingsMsg'
Learn More

Saccharin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015603 Supplier Merck Emd Millipore Supplier No. PHR13411G
Pharmaceutical (small molecule)

Specifications

CAS 81-07-2
Physical Form Neat
Linear Formula C7H5 NO3 S
MDL Number MFCD00005866
Melting Point 226°C to 229°C
Synonym Saccharin; 2,3-Dihydroxy-1,2-benzisothiazol-3-one-1,1-dioxide; 2-Sulfobenzoic acid imide; o-Benzoic sulfimide
Type Certified Reference Material
Quantity 1 g
Formula Weight 183.18
Grade Certified Reference Material