missing translation for 'onlineSavingsMsg'
Learn More

Sabinene hydrate, ≥97% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014569
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014569 500 mg
111014568 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014569 Supplier Merck Emd Millipore Supplier No. 96573500MGF

Sabinene hydrate is a monoterpene present in Origanum majorana, which is synthesized by the enzyme sabinene hydrate synthase.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 546-79-2
Physical Form Neat
Linear Formula C10H18 O
Melting Point 58°C to 62°C
Synonym 4-Thujanol
Type Analytical Standard
Quantity 500 mg
Formula Weight 154.25
Percent Purity ≥97% (GC)
Grade Analytical Standard