missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SAA Monoclonal Antibody (Reu86.5)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA140181

Catalog No. 01-676-543


Only null left
Explore available promotions

Description

Description

MA1-40181 detects SAA from human samples. MA1-40181 is raised against human SAA and Helix Pomatia Haemocyanine. It reacts specifically with SAA-1, the major isoform of SAA in plasma. MA1-40181 has been successfully used in immunohistochemistry (frozen and paraffin) applications. The MA1-40181 immunogen is human SAA.

The serum amyloid A family comprises a number of differentially expressed apolipoproteins, acute-phase SAA1 and SAA2, the former being the major component in plasma and constitutive SAAs. Although the liver is the primary site of synthesis of both SAA types extrahepatic production has been reported. The in vivo concentrations increase by as much as 1000 fold during inflammation. Several studies have expressed its importance in the diagnosis and monitoring of various diseases. Pathological SAA values are often detected in association with normal CRP concentrations; SAA rises earlier and more sharply than CRP. Recently, a broader view of SAA expression and function has been emerging.
Specifications

Specifications

SAA
Monoclonal
0.1 mg/mL
PBS with 0.1% BSA and 0.02% sodium azide
P0DJI8
SAA1
Human SAA.
100 μg
Primary
Human
Antibody
IgG1
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Reu86.5
Unconjugated
SAA1
amyloid a; Amyloid fibril protein AA; Amyloid protein A; MGC111216; PIG4; SAA; SAA1; Saa-1; Saa2; serum amyloid A; serum amyloid A 1; serum amyloid A 2; serum amyloid A protein; serum amyloid A1; Serum amyloid A1 protein; Serum amyloid A-1 protein; serum amyloid A-like protein; Serum amyloid protein A(2-102); Serum amyloid protein A(2-103); Serum amyloid protein A(2-104); Serum amyloid protein A(3-104); Serum amyloid protein A(4-101); Serum Amyloid-A; TP53I4; tumor protein p53 inducible protein 4
Mouse
Protein G
RUO
6288
4°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions