missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ S100Z Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149875
Description
S100Z Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Members of the S100 protein family contain 2 calcium-binding EF-hands and exhibit cell-type specific expression patterns. For additional background information on S100 proteins, see MIM 114085.Specifications
| S100Z | |
| Polyclonal | |
| Unconjugated | |
| S100Z | |
| Gm625; Protein S100-Z; S100 calcium binding protein Z; S100 calcium binding protein, zeta; S100 calcium-binding protein Z; S100Z; S100-zeta | |
| Rabbit | |
| Protein A | |
| RUO | |
| 100519888, 170591, 268686, 294661 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q8WXG8 | |
| S100Z | |
| Recombinant S100 Calcium Binding Protein Z (S100Z) (Met1-Lys99). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| Antibody |