missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ S100P Monoclonal Antibody (K40048_8D5), Invitrogen™
Mouse Monoclonal Antibody
$1,461.54 - $2,194.06
Specifications
| Antigen | S100P |
|---|---|
| Clone | K40048_8D5 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: MTELETAMGM IIDVFSRYSG SEGSTQTLTK GELKVLMEKE LPGFLQSGKD KDAVDKLLKD LDANGDAQVD FSEFIVFVAA ITSACHKYFE KAGLK
S100P Monoclonal antibody specifically detects S100P in Human samples. It is validated for Western BlotSpecifications
| S100P | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| MIG9; Migration-inducing gene 9 protein; protein S100-E; Protein S100-P; S100 calcium binding protein P; S100 calcium-binding protein P; S100E; S100p | |
| Recombinant human S100P (immunogen range is 1-95). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| S100P |
| K40048_8D5 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P25815 | |
| 6286 | |
| IgG1 | |
| Protein A | |
| Antibody |