missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ S100A1 Recombinant Rabbit Monoclonal Antibody (5L10L1)
GREENER_CHOICE

Catalog No. PIMA537861
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537861 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537861 Supplier Invitrogen™ Supplier No. MA537861
Only null left

Rabbit Recombinant Monoclonal Antibody

Positive Samples: Mouse lung, Mouse kidney, Mouse heart Immunogen sequence: GSELETAMET LINVFHAHSG KEGDKYKLSK KELKELLQTE LSGFLDAQKD VDAVDKVMKE LDENGDGEVD FQEYVVLVAA LTVACNNFFW ENS.

S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.

Specifications

Antigen S100A1
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 5L10L1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene S100A1
Gene Accession No. P56565
Gene Alias AI266795; Protein S100-A1; S100; S100 alpha; S100 calcium binding protein A1; S100 calcium-binding protein A1; S-100 protein alpha chain; S-100 protein subunit alpha; S100 protein, alpha polypeptide; S100a; S100A1; S100-alpha
Gene Symbols S100A1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 2-94 of human S100/S100A1 (P23297).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20193
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less