missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-Tetrahydrofurfurylamine 98.0+%, TCI America™

Supplier:  TCI America T2552200MG

 View more versions of this product

Catalog No. T2552200MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

(S)-(+)-Tetrahydrofurfurylamine
Yellow
200 mg
C5H11NO
2943
YNOGYQAEJGADFJ-YFKPBYRVSA-N
[(2S)-oxolan-2-yl]methanamine
641530
≥98.0% (GC,T)
7175-81-7
52°C
Clear Liquid at 20°C
MFCD00085303
s-+-tetrahydrofurfurylamine, s-tetrahydrofuran-2-yl methanamine, s-tetrahydrofurfurylamine, 2s-oxolan-2-ylmethanamine, 2s-oxolan-2-yl methanamine, 2-furanmethanamine, tetrahydro-, 2s, pubchem14101, pubchem17261, 2s-tetrahydrofurfurylamine
C1CC(OC1)CN
101.149
101.15

Chemical Identifiers

7175-81-7
101.149
YNOGYQAEJGADFJ-YFKPBYRVSA-N
641530
C1CC(OC1)CN
C5H11NO
MFCD00085303
s-+-tetrahydrofurfurylamine, s-tetrahydrofuran-2-yl methanamine, s-tetrahydrofurfurylamine, 2s-oxolan-2-ylmethanamine, 2s-oxolan-2-yl methanamine, 2-furanmethanamine, tetrahydro-, 2s, pubchem14101, pubchem17261, 2s-tetrahydrofurfurylamine
[(2S)-oxolan-2-yl]methanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions