missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-Tetrahydrofurfurylamine 98.0+%, TCI America™
$73.16 - $217.54
Chemical Identifiers
| CAS | 7175-81-7 |
|---|---|
| Molecular Formula | C5H11NO |
| Molecular Weight (g/mol) | 101.149 |
| MDL Number | MFCD00085303 |
| InChI Key | YNOGYQAEJGADFJ-YFKPBYRVSA-N |
| Synonym | s-+-tetrahydrofurfurylamine, s-tetrahydrofuran-2-yl methanamine, s-tetrahydrofurfurylamine, 2s-oxolan-2-ylmethanamine, 2s-oxolan-2-yl methanamine, 2-furanmethanamine, tetrahydro-, 2s, pubchem14101, pubchem17261, 2s-tetrahydrofurfurylamine |
| PubChem CID | 641530 |
| IUPAC Name | [(2S)-oxolan-2-yl]methanamine |
| SMILES | C1CC(OC1)CN |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T2552200MG
|
TCI America
T2552200MG |
200 mg |
Each for $73.16
|
|
|||||
|
T25521G
|
TCI America
T25521G |
1 g |
Each for $217.54
|
|
|||||
Chemical Identifiers
| 7175-81-7 | |
| 101.149 | |
| YNOGYQAEJGADFJ-YFKPBYRVSA-N | |
| 641530 | |
| C1CC(OC1)CN |
| C5H11NO | |
| MFCD00085303 | |
| s-+-tetrahydrofurfurylamine, s-tetrahydrofuran-2-yl methanamine, s-tetrahydrofurfurylamine, 2s-oxolan-2-ylmethanamine, 2s-oxolan-2-yl methanamine, 2-furanmethanamine, tetrahydro-, 2s, pubchem14101, pubchem17261, 2s-tetrahydrofurfurylamine | |
| [(2S)-oxolan-2-yl]methanamine |
Specifications
| 7175-81-7 | |
| 52°C | |
| Clear Liquid at 20°C | |
| MFCD00085303 | |
| s-+-tetrahydrofurfurylamine, s-tetrahydrofuran-2-yl methanamine, s-tetrahydrofurfurylamine, 2s-oxolan-2-ylmethanamine, 2s-oxolan-2-yl methanamine, 2-furanmethanamine, tetrahydro-, 2s, pubchem14101, pubchem17261, 2s-tetrahydrofurfurylamine | |
| C1CC(OC1)CN | |
| 101.149 | |
| 101.15 | |
| (S)-(+)-Tetrahydrofurfurylamine |
| Yellow | |
| 200 mg | |
| C5H11NO | |
| 2943 | |
| YNOGYQAEJGADFJ-YFKPBYRVSA-N | |
| [(2S)-oxolan-2-yl]methanamine | |
| 641530 | |
| ≥98.0% (GC,T) |
Safety and Handling
TSCA : No