missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Tetrahydrofuran-2-carboxylic Acid 98.0+%, TCI America™

Supplier:  TCI America T174125G

 View more versions of this product

Catalog No. T174125G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

(S)-(-)-Tetrahydrofuran-2-carboxylic Acid
Yellow
25 g
C5H8O3
3265
UJJLJRQIPMGXEZ-BYPYZUCNSA-N
(2S)-oxolane-2-carboxylic acid
446666
116.12
87392-07-2
251°C
Clear Liquid at 20°C
MFCD00272639
s-tetrahydrofuran-2-carboxylic acid, s---2-tetrahydrofuroic acid, s---tetrahydro-2-furoic acid, 2s-oxolane-2-carboxylic acid, s-tetrahydro-2-furoic acid, s---tetrahydrofuran-2-carboxylic acid, unii-0i2b46k2sj, tetrahydro-2-furoic acid,-, 2-furancarboxylic acid, tetrahydro-, 2s, s---2-carboxytetrahydrofuroic acid
C1CC(OC1)C(=O)O
116.116
CHEBI:45960
≥98.0% (GC,T)

Chemical Identifiers

87392-07-2
116.116
UJJLJRQIPMGXEZ-BYPYZUCNSA-N
446666
(2S)-oxolane-2-carboxylic acid
C5H8O3
MFCD00272639
s-tetrahydrofuran-2-carboxylic acid, s---2-tetrahydrofuroic acid, s---tetrahydro-2-furoic acid, 2s-oxolane-2-carboxylic acid, s-tetrahydro-2-furoic acid, s---tetrahydrofuran-2-carboxylic acid, unii-0i2b46k2sj, tetrahydro-2-furoic acid,-, 2-furancarboxylic acid, tetrahydro-, 2s, s---2-carboxytetrahydrofuroic acid
CHEBI:45960
C1CC(OC1)C(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions