missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-Tetrahydrofuran-2-carboxylic Acid 98.0+%, TCI America™
$29.83 - $369.08
Chemical Identifiers
| CAS | 87392-07-2 |
|---|---|
| Molecular Formula | C5H8O3 |
| Molecular Weight (g/mol) | 116.116 |
| MDL Number | MFCD00272639 |
| InChI Key | UJJLJRQIPMGXEZ-BYPYZUCNSA-N |
| Synonym | s-tetrahydrofuran-2-carboxylic acid, s---2-tetrahydrofuroic acid, s---tetrahydro-2-furoic acid, 2s-oxolane-2-carboxylic acid, s-tetrahydro-2-furoic acid, s---tetrahydrofuran-2-carboxylic acid, unii-0i2b46k2sj, tetrahydro-2-furoic acid,-, 2-furancarboxylic acid, tetrahydro-, 2s, s---2-carboxytetrahydrofuroic acid |
| PubChem CID | 446666 |
| ChEBI | CHEBI:45960 |
| IUPAC Name | (2S)-oxolane-2-carboxylic acid |
| SMILES | C1CC(OC1)C(=O)O |
Chemical Identifiers
| 87392-07-2 | |
| 116.116 | |
| UJJLJRQIPMGXEZ-BYPYZUCNSA-N | |
| 446666 | |
| (2S)-oxolane-2-carboxylic acid |
| C5H8O3 | |
| MFCD00272639 | |
| s-tetrahydrofuran-2-carboxylic acid, s---2-tetrahydrofuroic acid, s---tetrahydro-2-furoic acid, 2s-oxolane-2-carboxylic acid, s-tetrahydro-2-furoic acid, s---tetrahydrofuran-2-carboxylic acid, unii-0i2b46k2sj, tetrahydro-2-furoic acid,-, 2-furancarboxylic acid, tetrahydro-, 2s, s---2-carboxytetrahydrofuroic acid | |
| CHEBI:45960 | |
| C1CC(OC1)C(=O)O |
Specifications
| 87392-07-2 | |
| 251°C | |
| Clear Liquid at 20°C | |
| MFCD00272639 | |
| s-tetrahydrofuran-2-carboxylic acid, s---2-tetrahydrofuroic acid, s---tetrahydro-2-furoic acid, 2s-oxolane-2-carboxylic acid, s-tetrahydro-2-furoic acid, s---tetrahydrofuran-2-carboxylic acid, unii-0i2b46k2sj, tetrahydro-2-furoic acid,-, 2-furancarboxylic acid, tetrahydro-, 2s, s---2-carboxytetrahydrofuroic acid | |
| C1CC(OC1)C(=O)O | |
| 116.116 | |
| CHEBI:45960 | |
| ≥98.0% (GC,T) |
| Yellow | |
| 1 g | |
| C5H8O3 | |
| 3265 | |
| UJJLJRQIPMGXEZ-BYPYZUCNSA-N | |
| (2S)-oxolane-2-carboxylic acid | |
| 446666 | |
| 116.12 | |
| (S)-(-)-Tetrahydrofuran-2-carboxylic Acid |
Safety and Handling
TSCA : No