missing translation for 'onlineSavingsMsg'
Learn More

S-tert-Butyl-L-cysteine hydrochloride, 98%

CAS: 2481-09-6 | C7H16ClNO2S | 213.72 g/mol

$150.41 - $513.70

Chemical Identifiers

CAS 2481-09-6
MDL Number MFCD00037426
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5924406
View Documents
Thermo Scientific Chemicals
H5924406
5 g
Each for $150.41
Only null left
 
AAH5924414
View Documents
Thermo Scientific Chemicals
H5924414
25 g
Each for $513.70
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2481-09-6
MFCD00037426

Specifications

2481-09-6
C10H16Si2
1,4-bis dimethylsilyl benzene, benzene, 1,4-bis dimethylsilyl, p-phenylenebis dimethylsilane, 1,4-phenylenebis dimethylsilane, p-bis dimethylsilyl benzene, silane, p-phenylenebis dimethyl, silane, 1,4-phenylenebis dimethyl, 1,4 bis dimethylsilyl benzene
C[Si](=C1C=CC(=[Si](C)C)C=C1)C
5 g
6328729
98%
S-tert-Butyl-L-cysteine hydrochloride
White
MFCD00037426
UHXCHUWSQRLZJS-UHFFFAOYSA-N
(4-dimethylsilylidenecyclohexa-2,5-dien-1-ylidene)-dimethylsilane
192.41
213.73
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only