missing translation for 'onlineSavingsMsg'
Learn More

S-tert-Butyl-L-cysteine hydrochloride, 98%

Catalog No. aah5924414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5924414 25 g
AAH5924406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5924414 Supplier Thermo Scientific Chemicals Supplier No. H5924414
Only null left

CAS: 2481-09-6 | C7H16ClNO2S | 213.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2481-09-6
MDL Number MFCD00037426

Specifications

Color White
Molecular Formula C7H16ClNO2S
Synonym s-tert-butyl-l-cysteine hydrochloride, h-cys tbu-oh.hcl, h-cys tbu-oh hcl, s-t-butyl-l-cysteine hydrochloride, h-cys tbu-ohhcl, s-tert-butyl-l-cysteine-hydrogen chloride 1/1, 2r-2-amino-3-tert-butylsulfanyl propanoic acid hydrochloride, pubchem19026, h-cys but-oh hcl, h-cys but-oh hydrochloride
InChI Key MHBMYFJKEBCMDR-JEDNCBNOSA-N
SMILES CC(C)(C)SCC(C(=O)O)N.Cl
IUPAC Name (2R)-2-amino-3-tert-butylsulfanylpropanoic acid;hydrochloride
Quantity 25 g
Molecular Weight (g/mol) 213.72
PubChem CID 17750295
Formula Weight 213.73
Percent Purity 98%
Physical Form Powder
Chemical Name or Material S-tert-Butyl-L-cysteine hydrochloride
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only