missing translation for 'onlineSavingsMsg'
Learn More

S-Sodium Ethanethiosulfonate 98.0+%, TCI America™

Catalog No. S053325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S053325G 25 g
S0533500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S053325G Supplier TCI America Supplier No. S053325G

Chemical Identifiers

CAS 31999-88-9
Molecular Formula C2H6NaO2S2
Molecular Weight (g/mol) 149.178
InChI Key KVMDVKDQQJYYKE-UHFFFAOYSA-N
Synonym Ethanethiosulfonic Acid S-Sodium Salt
PubChem CID 87590989
IUPAC Name ethyl-hydroxy-oxo-sulfanylidene-$l^{6}-sulfane;sodium
SMILES CCS(=O)(=S)O.[Na]

Specifications

Color White
Quantity 25 g
Formula Weight 148.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material S-Sodium Ethanethiosulfonate