accessibility menu, dialog, popup

UserName

H3CSOSHONa

CAS RN 31999-88-9

IUPAC Name: 1-ethyl-1-hydroxy-1λ⁶-disulfen-1-one sodium
Synonyms: 1-ethyl-1-hydroxy-1λ⁶-disulfen-1-one sodium
Molecular Weight (g/mol): 149.18
Molecular Formula: C2H6NaO2S2
InChi Key: KVMDVKDQQJYYKE-UHFFFAOYNA-N
SMILES: [Na].CCS(O)(=O)=S
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)