missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-sec-Phenethyl alcohol, 99%

Catalog No. AC217790010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC217790010 1 g Glass bottle
AC217790050 5 g Glass bottle
AC217790250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC217790010 Supplier Thermo Scientific Chemicals Supplier No. 217790010

CAS: 1445-91-6 | C8H10O | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chiral derivatization reagent

Chemical Identifiers

CAS 1445-91-6
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00064264
InChI Key WAPNOHKVXSQRPX-ZETCQYMHSA-N
Synonym s-1-phenylethanol, s---1-phenylethanol, 1s-1-phenylethanol, 1s-1-phenylethan-1-ol, unii-2mic4qly2x, 1-phenylethanol, s, 2mic4qly2x, s-1-phenethyl alcohol, s-1-phenyl-1-ethanol, s-1-phenylethan-1-ol
PubChem CID 443135
ChEBI CHEBI:16346
SMILES C[C@H](O)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 9°C to 11°C
Color Colorless
Density 1.0100g/mL
Boiling Point 98°C (20.0 mmHg)
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5260 to 1.528
Linear Formula C6H5CH(CH3)OH
Quantity 1 g
Beilstein 06, IV, 3029
Specific Gravity 1.01
Specific Rotation Condition −42.50 (20.00°C neat)
Specific Rotation -42.5
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in glycerol
IUPAC Name (1S)-1-phenylethanol
Formula Weight 122.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material (S)-(-)-sec-Phenethyl alcohol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only